Assay Details
| Gene Symbol: Mmrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751550.3 | XP_003751598.2 | 5 - 6 | 812 | 69 | |
| GenBank mRNA | FQ218167.1 | - | 5 - 6 | 747 | 69 | |
More Information
Set Membership: |
|

Mmrn2
Genomic Map