Assay Details
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751115.3 | XP_003751163.1 | 946 | 95 | ||
| XM_341029.6 | XP_341030.3 | 936 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751115.3 | XP_003751163.1 | 946 | 95 | ||
| XM_341029.6 | XP_341030.3 | 936 | 95 | |||
Set Membership: |
|