Assay Details
| Gene Symbol: Snx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024999.1 | NP_001020170.1 | 1 - 2 | 95 | 67 | |
| GenBank mRNA | BC091194.1 | - | 1 - 2 | 95 | 7373920 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC091194.1, NM_001024999.1 transcripts.
Set Membership: |
|

Snx20
Genomic Map