Assay Details
| Gene Symbol: Ankrd34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109174.1 | NP_001102644.1 | 3 - 4 | 540 | 84 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109174.1 transcripts.
Set Membership: |
|

Ankrd34b
Genomic Map