Assay Details
| Gene Symbol: RGD1306233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106564.1 | NP_001100034.1 | 1 - 2 | 211 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306233| Gene Symbol: RGD1306233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106564.1 | NP_001100034.1 | 1 - 2 | 211 | 82 | |
Set Membership: |
|