Assay Details
| Gene Symbol: Prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053576.2 | NP_446028.1 | 1 - 2 | 114 | 58 | |
| GenBank mRNA | AF014009.1 | - | 1 - 2 | 114 | 58 | |
| Y17295.1 | - | 128 | 58 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prdx6
Genomic Map