Assay Details
| Gene Symbol: Cdc14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108404.1 | NP_001101874.1 | 9 - 10 | 1131 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
biological_process
peptidyl-tyrosine dephosphorylation
positive regulation of ubiquitin protein ligase activity
molecular_function
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein dephosphorylation
biological_process
peptidyl-tyrosine dephosphorylation
positive regulation of ubiquitin protein ligase activity
molecular_function
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P134435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34256 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134435 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134435 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P134435 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17930 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6502 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93785 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10793476 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793479 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793487 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793488 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793491 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793480 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793478 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793485 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10793475 | - | - |
| Rat Genome 230 2.0 Array | 1392602_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392602_at | - | - |
| Affymetrix HT Rat Focus Array | 1392602_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392602_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700008172.rn.1 | TC1700001185.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700008173.rn.1 | TC1700001185.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392602_at | - | - |
| Rat Expression Set 230 | 1392602_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793475 | - | - |
| Rat Genome U34 Set | rc_AI232772_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21990429 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355439 | - | - |

Cdc14b
Genomic Map