Assay Details
| Gene Symbol: Abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231162.2 | XP_006231224.1 | 1 - 2 | 371 | 146 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006231162.2 transcripts.
Set Membership: |
|

Abhd17b
Genomic Map