Assay Details
| Gene Symbol: Gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013151.1 | NP_001013169.1 | 6 - 7 | 1364 | 150 | |
| XM_008760288.1 | XP_008758510.1 | 7 - 8 | 2001 | 150 | ||
| GenBank mRNA | BC090316.1 | - | 6 - 7 | 1364 | 7305112 | 150 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
biological_process
metabolic process
phospholipase C-activating dopamine receptor signaling pathway
G-protein coupled receptor binding
molecular_function
GTPase activity
signal transducer activity
GTP binding
G-protein beta/gamma-subunit complex binding
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
biological_process
metabolic process
phospholipase C-activating dopamine receptor signaling pathway
G-protein coupled receptor binding
molecular_function
GTPase activity
signal transducer activity
GTP binding
G-protein beta/gamma-subunit complex binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P593161 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P377396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100604 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P377396 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P593161 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P593161 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P377396 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P377396 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P593161 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P377396 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P593161 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77033 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101972 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87034 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10714219 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714215 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714216 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714217 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10714214 | - | - |
| Rat Genome 230 2.0 Array | 1393825_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381557_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381557_at | - | - |
| Rat Genome 230 2.0 Array | 1393825_at | - | - |
| Affymetrix HT Rat Focus Array | 1381557_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393825_at | - | - |
| Affymetrix HT Rat Focus Array | 1393825_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381557_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100027150.rn.1 | TC0100003324.rn.1 | RTA TC |
| Rat Expression Set 230 | 1381557_at | - | - |
| Rat Expression Set 230 | 1393825_PM_at | - | - |
| Rat Expression Set 230 | 1381557_PM_at | - | - |
| Rat Expression Set 230 | 1393825_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22375458 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650309 | - | - |

Gna14
Genomic Map