Assay Details
| Gene Symbol: Slc41a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108742.1 | NP_001102212.1 | 9 - 10 | 1763 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130983 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P878617 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P878617 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P878617 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P321544 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P108304 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99894 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69011 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10894609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894614 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894608 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10894606 | - | - |
| Rat Genome 230 2.0 Array | 1384934_at | - | - |
| Rat Genome 230 2.0 Array | 1379805_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379805_at | - | - |
| Rat Genome 230 2.0 Array | 1384934_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379805_at | - | - |
| Affymetrix HT Rat Focus Array | 1379805_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384934_at | - | - |
| Affymetrix HT Rat Focus Array | 1384934_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700003621.rn.1 | TC0700000419.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700003623.rn.1 | TC0700000419.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700003622.rn.1 | TC0700000419.rn.1 | RTA TC |
| Rat Expression Set 230 | 1384934_PM_at | - | - |
| Rat Expression Set 230 | 1384934_at | - | - |
| Rat Expression Set 230 | 1379805_at | - | - |
| Rat Expression Set 230 | 1379805_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894606 | - | - |
| Rat Genome U34 Set | rc_AA893454_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21451109 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350462 | - | - |

Slc41a2
Genomic Map