Assay Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024896.1 | NP_001020067.1 | 2 - 3 | 986 | 93 | |
| XM_006255712.2 | XP_006255774.1 | 3 - 4 | 970 | 93 | ||
| XM_006255713.2 | XP_006255775.1 | 3 - 4 | 763 | 93 | ||
| XM_006255714.2 | XP_006255776.1 | 3 - 4 | 1923 | 93 | ||
| XM_008772622.1 | XP_008770844.1 | 4 - 5 | 985 | 93 | ||
| GenBank mRNA | BC097457.1 | - | 2 - 3 | 986 | 7457192 | 93 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hsdl1
Genomic Map