Assay Details
| Gene Symbol: Nfat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107425.1 | NP_001100895.1 | 5 - 6 | 1808 | 65 | |
| XM_003751872.3 | XP_003751920.1 | 4 - 5 | 1469 | 65 | ||
| XM_006255553.2 | XP_006255615.1 | 3 - 4 | 4231 | 65 | ||
| XM_006255554.2 | XP_006255616.1 | 3 - 4 | 3812 | 65 | ||
| XM_006255555.2 | XP_006255617.1 | 4 - 5 | 479 | 65 | ||
| XM_006255556.2 | XP_006255618.1 | 3 - 4 | 416 | 65 | ||
| XM_008772503.1 | XP_008770725.1 | 5 - 6 | 1523 | 65 | ||
| XM_008772504.1 | XP_008770726.1 | 3 - 4 | 1350 | 65 | ||
| XM_008772505.1 | XP_008770727.1 | 2 - 3 | 4264 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
transcription, DNA-templated
response to osmotic stress
positive regulation of gene expression
positive regulation of transcription from RNA polymerase II promoter
regulation of calcineurin-NFAT signaling cascade
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
cytokine production
transcription, DNA-templated
response to osmotic stress
positive regulation of gene expression
positive regulation of transcription from RNA polymerase II promoter
regulation of calcineurin-NFAT signaling cascade
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P555795 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137375 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P555795 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21944 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21944 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P555795 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21944 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P555795 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P431447 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19036 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24306 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70247 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10807649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807650 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807651 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807655 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807653 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807646 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807652 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807654 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10807638 | - | - |
| Rat Genome 230 2.0 Array | 1398421_at | - | - |
| Rat Genome 230 2.0 Array | 1398421_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372772_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372772_at | - | - |
| Affymetrix HT Rat Focus Array | 1372772_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372772_at | - | - |
| Affymetrix HT Rat Focus Array | 1398421_at | - | - |
| Affymetrix HT Rat Focus Array | 1398421_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900011257.rn.1 | TC1900001379.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398421_PM_at | - | - |
| Rat Expression Set 230 | 1372772_at | - | - |
| Rat Expression Set 230 | 1372772_PM_at | - | - |
| Rat Expression Set 230 | 1398421_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807638 | - | - |
| Rat Genome U34 Set | rc_AI177168_at | - | - |
| Rat Genome U34 Set | rc_AI230823_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20869337 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373056 | - | - |

Nfat5
Genomic Map