Assay Details
| Gene Symbol: Mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010947.1 | NP_001010947.1 | 2 - 3 | 588 | 89 | |
| GenBank mRNA | BC089060.1 | - | 2 - 3 | 588 | 7320396 | 89 |
| FQ213644.1 | - | 2 - 3 | 580 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mri1
Genomic Map