Assay Details
| Gene Symbol: Fcgr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100836.1 | NP_001094306.1 | 1 - 2 | 48 | 67 | |
| GenBank mRNA | BC158809.1 | - | 1 - 2 | 49 | 7364883 | 67 |
| CO555980.1 | - | 1 - 2 | 81 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcgr1a
Genomic Map