Assay Details
| Gene Symbol: Fcgr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100836.1 | NP_001094306.1 | 3 - 4 | 323 | 87 | |
| GenBank mRNA | BC158809.1 | - | 3 - 4 | 324 | 7364883 | 87 |
| CO555980.1 | - | 3 - 4 | 356 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcgr1a
Genomic Map