Assay Details
| Gene Symbol: Nr1h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749366.3 | XP_003749414.1 | 3 - 4 | 620 | 99 | |
| XM_008761431.1 | XP_008759653.1 | 3 - 4 | 624 | 99 | ||
More Information
Set Membership: |
|

Nr1h5
Genomic Map