Assay Details
| Gene Symbol: Dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252176.2 | XP_006252238.1 | 1 - 2 | 1020 | 106 | |
| GenBank mRNA | BC158600.1 | - | 1 - 2 | 537 | 7130710 | 106 |
More Information
Updated Mapping Information
Set Membership: |
|

Dleu7
Genomic Map