Assay Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107861.1 | NP_001101331.1 | 1 - 2 | 72 | 111 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107861.1 transcripts.
Set Membership: |
|

Kbtbd2
Genomic Map