Assay Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025015.1 | NP_001020186.1 | 1 - 2 | 357 | 138 | |
| GenBank mRNA | BC091236.1 | - | 1 - 2 | 357 | 7322967 | 138 |
| FQ216550.1 | - | 1 - 2 | 356 | 138 | ||
More Information
Set Membership: |
|

Gtpbp8
Genomic Map