Assay Details
| Gene Symbol: Fam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107102.1 | NP_001100572.1 | 3 - 4 | 384 | 70 | |
| XM_006248157.2 | XP_006248219.1 | 3 - 4 | 447 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam3b
Genomic Map