Assay Details
| Gene Symbol: St6gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100888.1 | NP_001094358.1 | 5 - 6 | 1346 | 98 | |
| XM_006244260.2 | XP_006244322.1 | 4 - 5 | 1551 | 98 | ||
| XM_006244264.2 | XP_006244326.1 | 4 - 5 | 1414 | 98 | ||
| XM_006244265.2 | XP_006244327.1 | 4 - 5 | 1718 | 98 | ||
| XM_008766756.1 | XP_008764978.1 | 4 - 5 | 3521 | 98 | ||
| XM_008766757.1 | XP_008764979.1 | 4 - 5 | 1804 | 98 | ||
| XM_008766758.1 | XP_008764980.1 | 4 - 5 | 3521 | 98 | ||
| GenBank mRNA | AJ627626.1 | - | 3 - 4 | 1124 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

St6gal2
Genomic Map