Assay Details
| Gene Symbol: Pak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106238.1 | NP_001099708.1 | 5 - 6 | 1343 | 66 | |
| XM_008759119.1 | XP_008757341.1 | 5 - 6 | 3393 | 66 | ||
| GenBank mRNA | BC168940.1 | - | 5 - 6 | 1353 | 8363273 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P067105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86168 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067103 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P547036 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P547036 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067103 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P067103 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P547036 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21423 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7394 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10720253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720254 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720255 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10720247 | - | - |
| Rat Genome 230 2.0 Array | 1378380_at | - | - |
| Rat Genome 230 2.0 Array | 1396426_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378380_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396426_at | - | - |
| Affymetrix HT Rat Focus Array | 1378380_at | - | - |
| Affymetrix HT Rat Focus Array | 1378380_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100065139.rn.1 | TC0100008032.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100065141.rn.1 | TC0100008032.rn.1 | RTA TC |
| Rat Expression Set 230 | 1378380_at | - | - |
| Rat Expression Set 230 | 1396426_PM_at | - | - |
| Rat Expression Set 230 | 1378380_PM_at | - | - |
| Rat Expression Set 230 | 1396426_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10720247 | - | - |
| Rat Genome U34 Set | rc_AI030619_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20756373 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359465 | - | - |

Pak4
Genomic Map