Assay Details
| Gene Symbol: Zfp112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107487.1 | NP_001100957.1 | 2 - 3 | 229 | 73 | |
| XM_006228422.2 | XP_006228484.1 | 3 - 4 | 393 | 73 | ||
| XM_008758926.1 | XP_008757148.1 | 2 - 3 | 253 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp112
Genomic Map