Assay Details
| Gene Symbol: Fut9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053465.1 | NP_445917.1 | 1 - 2 | 251 | 72 | |
| GenBank mRNA | AB049819.1 | - | 1 - 2 | 251 | 72 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB049819.1,NM_053465.1 transcripts.
Set Membership: |
|

Fut9
Genomic Map