Assay Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025034.1 | NP_001020205.1 | 3 - 4 | 466 | 93 | |
| XM_008771879.1 | XP_008770101.1 | 3 - 4 | 865 | 93 | ||
| XM_008771880.1 | XP_008770102.1 | 3 - 4 | 488 | 93 | ||
| XM_008771881.1 | XP_008770103.1 | 3 - 4 | 914 | 93 | ||
| XM_008771882.1 | XP_008770104.1 | 3 - 4 | 501 | 93 | ||
| XM_008771883.1 | XP_008770105.1 | 3 - 4 | 1118 | 93 | ||
| XM_008771884.1 | XP_008770106.1 | 3 - 4 | 510 | 93 | ||
| GenBank mRNA | BC085116.1 | - | 3 - 4 | 466 | 7107468 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P759788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24330 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P759788 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P759788 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P759788 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P748575 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P829888 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P759788 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6451 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13169 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83273 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91168 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31298 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103825 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10799619 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799616 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10799615 | - | - |
| Rat Genome 230 2.0 Array | 1382044_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389146_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389146_at | - | - |
| Rat Genome 230 2.0 Array | 1382044_at | - | - |
| Affymetrix HT Rat Focus Array | 1382044_at | - | - |
| Affymetrix HT Rat Focus Array | 1389146_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389146_at | - | - |
| Affymetrix HT Rat Focus Array | 1382044_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700014872.rn.1 | TC1700002235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014871.rn.1 | TC1700002235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014875.rn.1 | TC1700002235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014870.rn.1 | TC1700002235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014867.rn.1 | TC1700002235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014884.rn.1 | TC1700002235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700014869.rn.1 | TC1700002235.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389146_PM_at | - | - |
| Rat Expression Set 230 | 1389146_at | - | - |
| Rat Expression Set 230 | 1382044_PM_at | - | - |
| Rat Expression Set 230 | 1382044_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10799615 | - | - |
| Rat Genome U34 Set | rc_AA943508_at | - | - |
| Rat Genome U34 Set | rc_AA818780_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21052517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360024 | - | - |

Fam107b
Genomic Map