Assay Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107866.2 | NP_001101336.1 | 2 - 3 | 464 | 65 | |
| XM_008763023.1 | XP_008761245.1 | 2 - 3 | 1014 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Loxl3
Genomic Map