Assay Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172331.1 | NP_758834.1 | 2 - 3 | 124 | 113 | |
| GenBank mRNA | AY150169.1 | - | 2 - 3 | 124 | 113 | |
| FQ215758.1 | - | 2 - 3 | 138 | 113 | ||
| FQ219218.1 | - | 2 - 3 | 143 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chmp3
Genomic Map