Assay Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107444.1 | NP_001100914.1 | 2 - 3 | 749 | 57 | |
| XM_006231645.2 | XP_006231707.1 | 2 - 3 | 749 | 57 | ||
| XM_008760536.1 | XP_008758758.1 | 2 - 3 | 523 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nhlrc2
Genomic Map