Assay Details
| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181382.2 | NP_852047.1 | 4 - 5 | 730 | 87 | |
| GenBank mRNA | AY266116.1 | - | 4 - 5 | 730 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gldn
Genomic Map