Assay Details
Gene Symbol: Pts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017220.1 | NP_058916.1 | 4 - 5 | 296 | 100 | |
GenBank mRNA | BC059140.1 | - | 4 - 5 | 279 | 6888665 | 100 |
M77850.1 | - | 4 - 5 | 296 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
