Assay Details
| Gene Symbol: Siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108759.1 | NP_001102229.1 | 3 - 4 | 447 | 67 | |
| GenBank mRNA | BC162050.1 | - | 3 - 4 | 418 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siae
Genomic Map