Assay Details
| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012117.1 | NP_001012117.2 | 2 - 3 | 1717 | 118 | |
| XM_006241535.2 | XP_006241597.1 | 2 - 3 | 1475 | 118 | ||
| GenBank mRNA | BC082107.1 | - | 2 - 3 | 1717 | 7134870 | 118 |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo43
Genomic Map