Assay Details
| Gene Symbol: Rrp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130568.1 | NP_001124040.1 | 3 - 4 | 392 | 78 | |
| GenBank mRNA | BC079405.1 | - | 3 - 4 | 347 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrp7a
Genomic Map