Assay Details
| Gene Symbol: Cmtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014031.1 | NP_001014053.1 | 20 - 21 | 2194 | 108 | |
| XM_008772816.1 | XP_008771038.1 | 20 - 21 | 2264 | 108 | ||
| GenBank mRNA | AY539880.1 | - | 2438 | 108 | ||
| BC085799.1 | - | 20 - 21 | 2194 | 7110279 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P095675 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P196933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159023 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P095675 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P196933 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P196933 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P095675 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P095675 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P196933 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P196933 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15057 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12085 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57451 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62916 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10828935 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828931 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828932 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828937 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828933 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828934 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828939 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828918 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828943 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828938 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828936 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828927 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828942 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828945 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828920 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10828913 | - | - |
| Rat Genome 230 2.0 Array | 1398925_at | - | - |
| Rat Genome 230 2.0 Array | 1398925_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398925_at | - | - |
| Affymetrix HT Rat Focus Array | 1398925_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000003046.rn.1 | TC2000000323.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398925_at | - | - |
| Rat Expression Set 230 | 1398925_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828913 | - | - |
| Rat Genome U34 Set | rc_AI113020_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650366 | - | - |

Cmtr1
Genomic Map