Assay Details
| Gene Symbol: Plekhg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240253.2 | XP_006240315.1 | 11 - 12 | 3400 | 61 | |
| XM_006240254.2 | XP_006240316.1 | 11 - 12 | 3400 | 61 | ||
| XM_008764865.1 | XP_008763087.1 | 11 - 12 | 1794 | 61 | ||
| XM_008764866.1 | XP_008763088.1 | 9 - 10 | 1266 | 61 | ||
More Information
Set Membership: |
|

Plekhg3
Genomic Map