Assay Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017469.1 | NP_001017469.1 | 4 - 5 | 528 | 126 | |
| GenBank mRNA | BC078980.1 | - | 4 - 5 | 528 | 7112035 | 126 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam71d
Genomic Map