Assay Details
| Gene Symbol: Wdr35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099340.1 | NP_001092810.1 | 10 - 11 | 1298 | 87 | |
| GenBank mRNA | EF613262.1 | - | 10 - 11 | 1298 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr35l
Genomic Map