Assay Details
| Gene Symbol: Wdr35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099340.1 | NP_001092810.1 | 1 - 2 | 124 | 101 | |
| XM_006239885.2 | XP_006239947.1 | 1 - 2 | 140 | 101 | ||
| GenBank mRNA | EF613262.1 | - | 1 - 2 | 124 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the EF613262.1, NM_001099340.1, XM_006239885.2 transcripts.
Set Membership: |
|

Wdr35l
Genomic Map