Assay Details
| Gene Symbol: Ndn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008558.1 | NP_001008558.1 | 1 | 222 | 89 | |
| GenBank mRNA | BC087630.1 | - | 1 | 222 | 7319134 | 89 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndn
Genomic Map