Assay Details
| Gene Symbol: Olr1522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001000040.1 | NP_001000040.1 | 1 | 887 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1522| Gene Symbol: Olr1522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001000040.1 | NP_001000040.1 | 1 | 887 | 82 | |
Set Membership: |
|