Assay Details
| Gene Symbol: Ddx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199119.1 | NP_954550.1 | 5 - 6 | 1946 | 78 | |
| XM_006240483.1 | XP_006240545.1 | 5 - 6 | 2149 | 78 | ||
| XM_006240484.2 | XP_006240546.1 | 5 - 6 | 2381 | 78 | ||
| GenBank mRNA | AY341876.1 | - | 5 - 6 | 1946 | 78 | |
| BC097262.1 | - | 5 - 6 | 1933 | 7442976 | 78 | |
| FQ212095.1 | - | 3 - 4 | 692 | 78 | ||
| FQ218902.1 | - | 1 - 2 | 63 | 78 | ||
| Gene Symbol: LOC100909476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750226.3 | XP_003750274.3 | 2 - 3 | 1275 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx24
Genomic Map