Assay Details
| Gene Symbol: Amigo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206881.1 | NP_996764.1 | 1 | 149 | 70 | |
| XM_006233140.2 | XP_006233202.1 | 2 | 549 | 70 | ||
| XM_008761391.1 | XP_008759613.1 | 2 | 584 | 70 | ||
| XM_008761392.1 | XP_008759614.1 | 1 | 1006 | 70 | ||
| GenBank mRNA | AY237729.1 | - | 1 | 149 | 70 | |
| BC167749.1 | - | 1 | 193 | 9021924 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Amigo1
Genomic Map