Assay Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213561.3 | NP_998726.2 | 596 | 61 | ||
| GenBank mRNA | BC093374.1 | - | 523 | 7388599 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tcf19
Genomic Map