Assay Details
| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107257.1 | NP_001100727.1 | 2669 | 150 | ||
| GenBank mRNA | BC168877.1 | - | 2826 | 7134856 | 150 | |
| FQ212387.1 | - | 2743 | 150 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P379878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139983 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P379878 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P379878 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P379878 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P379878 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3818 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104720 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10779680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779679 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10779678 | - | - |
| Rat Genome 230 2.0 Array | 1372662_at | - | - |
| Rat Genome 230 2.0 Array | 1372662_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372662_at | - | - |
| Affymetrix HT Rat Focus Array | 1372662_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500001733.rn.1 | TC1500000245.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500001732.rn.1 | TC1500000245.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500001724.rn.1 | TC1500000245.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500001727.rn.1 | TC1500000245.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372662_at | - | - |
| Rat Expression Set 230 | 1372662_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779678 | - | - |
| Rat Genome U34 Set | rc_AI176698_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21639170 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357596 | - | - |

Fbxo34
Genomic Map