Assay Details
| Gene Symbol: Irf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108859.1 | NP_001102329.1 | 8 | 3952 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle arrest
biological_process
negative regulation of cell proliferation
keratinocyte differentiation
skin development
keratinocyte proliferation
positive regulation of transcription, DNA-templated
cell development
mammary gland epithelial cell differentiation
regulatory region DNA binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
cell cycle arrest
biological_process
negative regulation of cell proliferation
keratinocyte differentiation
skin development
keratinocyte proliferation
positive regulation of transcription, DNA-templated
cell development
mammary gland epithelial cell differentiation
regulatory region DNA binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P638014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148115 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P638014 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P638014 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P638014 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P638014 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P215326 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31715 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10766803 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766799 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766798 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766802 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766804 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766800 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766801 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766805 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10766797 | - | - |
| Rat Genome 230 2.0 Array | 1377379_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377379_at | - | - |
| Rat Genome 230 2.0 Array | 1386568_at | - | - |
| Rat Genome 230 2.0 Array | 1386568_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377379_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377379_at | - | - |
| Affymetrix HT Rat Focus Array | 1386568_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386568_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300007627.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007620.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007623.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007625.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007626.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007624.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007622.rn.1 | TC1300001078.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007621.rn.1 | TC1300001078.rn.1 | RTA TC |
| Rat Expression Set 230 | 1386568_PM_at | - | - |
| Rat Expression Set 230 | 1377379_at | - | - |
| Rat Expression Set 230 | 1377379_PM_at | - | - |
| Rat Expression Set 230 | 1386568_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766797 | - | - |
| Rat Genome U34 Set | rc_AA996727_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22384771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356810 | - | - |

Irf6
Genomic Map