Assay Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240439.2 | XP_006240501.1 | 15 | 3499 | 121 | |
| XM_006240441.2 | XP_006240503.1 | 15 | 3560 | 121 | ||
| XM_006240442.2 | XP_006240504.1 | 13 | 4312 | 121 | ||
| XM_008764817.1 | XP_008763039.1 | 15 | 3554 | 121 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1311756
Genomic Map