Assay Details
| Gene Symbol: LOC100125364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103356.2 | NP_001096826.1 | 2 | 2413 | 127 | |
| XM_006227969.2 | XP_006228031.1 | 2 | 2346 | 127 | ||
| XM_006227970.2 | XP_006228032.1 | 2 | 2324 | 127 | ||
| XM_008758746.1 | XP_008756968.1 | 2 | 2419 | 127 | ||
| GenBank mRNA | BG373545.1 | - | 1 | 127 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100125364
Genomic Map