Assay Details
| Gene Symbol: Spred2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047094.1 | NP_001040559.1 | 355 | 145 | ||
| GenBank mRNA | AB125136.1 | - | 355 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
multicellular organism development
negative regulation of peptidyl-threonine phosphorylation
positive regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of protein serine/threonine kinase activity
regulation of protein deacetylation
stem cell factor receptor binding
protein kinase binding
protein serine/threonine kinase inhibitor activity
inactivation of MAPK activity
multicellular organism development
negative regulation of peptidyl-threonine phosphorylation
positive regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of protein serine/threonine kinase activity
regulation of protein deacetylation
stem cell factor receptor binding
protein kinase binding
protein serine/threonine kinase inhibitor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P163652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2070 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P163652 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P163652 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P163652 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P163652 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38620 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64575 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38308 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95521 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94552 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39385 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10774348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774352 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10774345 | - | - |
| Rat Genome 230 2.0 Array | 1389008_at | - | - |
| Rat Genome 230 2.0 Array | 1389008_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400007777.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007784.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007778.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007780.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007781.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007779.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007782.rn.1 | TC1400001079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007783.rn.1 | TC1400001079.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389008_at | - | - |
| Rat Expression Set 230 | 1389008_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774345 | - | - |
| Rat Genome U34 Set | rc_AA943506_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371095 | - | - |

Spred2
Genomic Map