Assay Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033900.2 | NP_001029072.2 | 3 | 1043 | 126 | |
| NR_028055.1 | 2 | 1422 | 126 | |||
| GenBank mRNA | BC101926.1 | - | 3 | 1017 | 7939318 | 126 |
| FQ215222.1 | - | 3 | 1046 | 126 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ngrn
Genomic Map