Assay Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022175.1 | NP_071511.1 | 1 | 42 | 83 | |
| GenBank mRNA | DQ058653.1 | - | 1 | 42 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rhox5
Genomic Map